missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Agar Irradiated (Triple Wrap) Plate

Product Code. 12912618
Click to view available options
Format:
140mm Plate
Color:
Straw, clear
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 12912618

Brand: Thermo Scientific™ PO0306T

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Carry out environmental monitoring in the most regulated environments with this Triple Wrapped Plate for passive and active air sampling applications.

Experience next-level cleanroom confidence with Thermo Scientific™ Oxoid™ Tryptone Soya Agar Irradiated Plate. The Tryptone Soya Agar supports the growth of wide variety of organisms.

Perform effortless environmental monitoring with Tryptone Soya Agar Irradiated (Triple Wrap) Plate.

  • Sterile: plates are irradiated and triple wrapped in a transparent packaging.
  • Consistent: automated manufacturing procedures ensure a smooth, evenly distributed layer of agar on each plate.
  • Optimized: products undergo quantitative quality control testing, using a low dose inoculum to ensure optimum performance.
  • Ready-to-use: convinince of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, clear
Content And Storage 2 to 25°C
Description Tryptone Soya Agar Irradiated Triple Wrap
Format 140mm Plate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 88.0g – 95.0g
Packaging Type Triple Wrapped - Irradiated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.