Learn More
Thermo Scientific™ Tryptone Soya Agar, TWI, Deep Fill
Descripción
Monitor aseptic processes within manufacturing cleanrooms or isolators with Thermo Scientific™ Tryptone Soya Agar (TWI, Deep Fill). A general purpose agar medium, containing two peptones, Tryptone Soya Agar (TWI, Deep Fill) will support the growth of a wide variety of organisms. It is suitable for the cultivation both of aerobes and anaerobes.
- Sterile: plates are irradiated and triple-wrapped in transparent packaging
- Consistent: automated manufacturing procedures ensure a smooth, evenly distributed layer of agar on each plate
- Optimized: products undergo quantitative quality control testing, using a low-dose inoculum to ensure optimum performance
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificaciones
Especificaciones
| Content And Storage | 2° to 25°C |
| Description | Tryptone Soya Agar (TWI, Deep Fill) |
| Format | 90mm Monoplate |
| Product Type | Agar Plate |
| Fill Volume | 23 mL |
| Quantity | 1 x 10 Plates |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.