missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Tryptone Soya Broth (Bottled)

Product Code. 13255489
Change view
Click to view available options
Fill Weight:
10.01 to 0.3 g
10.0g + 0.3g
100.0 - 101.5g
100.0g + 1.5 g
20.0 - 20.3 g
500.0 - 504.0 g
90 to 90.5 g
Unit Size:
Pack of 10
Pack of 24
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Unit Size
13255489 500.0 - 504.0 g Pack of 10
12786959 10.0g + 0.3g Pack of 24
12693428 100.0g + 1.5 g Pack of 10
11983112 10.01 to 0.3 g Pack of 24
15239047 20.0 - 20.3 g Pack of 24
12864132 90 to 90.5 g Pack of 10
12854132 100.0 - 101.5g Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 13255489 Supplier Thermo Scientific™ Supplier No. BO0351V
  • ONLINE EXCLUSIVE PRICE
    185.93 €
    173.36 € / Pack of 10
    Save 12.57 €
    6% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grow a wide variety of microprganisms using this medium in convenient ready prepared bottles.

Grow a wide variety of microorganisms with Thermo Scientific™ Tryptone Soya Broth ready-prepared bottled medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Tryptone Soya Broth.

  • Conforms to formulations detailed in International Pharmacopoeia.
  • Convenient: pre-filled bottles are ready for immediate use.
  • Simple: easy to use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, clear
Form Liquid
Manufacturing Origin United Kingdom
Product Type Broth Tube
Sterility Sterile
Target Organism Class Bacteria
Test Type Microbial Detection
Fill Weight 500.0 - 504.0 g
Quantity 10 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.