missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Tryptone Soya Broth, Wide Neck

Product Code. 10239999
Change view
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10239999 10 x 500 mL Pack of 10
12573936 10 x 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10239999 Supplier Thermo Scientific™ Supplier No. BO0509V
  • ONLINE EXCLUSIVE PRICE
    161.00 €
    155.45 € / Pack of 10
    Save 5.55 €
    3% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wide neck bottle with septum

General purpose bottled media complying with EP/USP specification.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® ARTG
Certifications/Compliance EP/USP
Content And Storage 2°C to 25°C
Country of Origin UK
Media Type Prepared Media
Product Type Microbiology Media
For Use With (Application) Pharmaceutical Bottled Media
Packaging Type Pk.
Quantity 10 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.