Learn More
TSC22D4 Monoclonal Antibody (OTI3G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811733S
Encompass_Preferred
 View more versions of this product
Product Code. 17525944
  • 242.00 € / 30µL
Description
TSC22D4 Monoclonal antibody specifically detects TSC22D4 in Human samples. It is validated for Western Blot
Specifications
Specifications
TSC22D4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P00742
Mouse
Affinity chromatography
RUO
2159
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3G6
Unconjugated
F10
0610009M14Rik; 1700023B23Rik; AI415410; LOW QUALITY PROTEIN: TSC22 domain family protein 4; THG1; THG-1; Thg1pit; Thg-1pit; Tilz2; TSC22 domain family 4; TSC22 domain family member 4; TSC22 domain family protein 4; TSC22 domain family, member 4; Tsc22d4; tsc-22-like protein THG-1; TSC22-related inducible leucine zipper 2; TSC22-related-inducible leucine zipper protein 2
Human recombinant protein fragment corresponding to amino acids 41-488 of human F10 produced in E.coli.
30 μL