missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC22D4 Monoclonal Antibody (OTI3G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811733
This item is not returnable.
View return policy
Description
TSC22D4 is a leucine zipper-containing protein that is highly conserved during evolution. It is transcriptionally up-regulated by many different stimuli, including anti-cancer drugs and growth inhibitors. TSC22D4 may play a suppressive role in tumorigenesis.Specifications
| TSC22D4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y3Q8 | |
| TSC22D4 | |
| Human recombit protein fragment corresponding to amino acids 1-395 of human TSC22D4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3G6 | |
| Unconjugated | |
| TSC22D4 | |
| THG-1, THG1, TILZ2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 81628 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction