missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSG Monoclonal Antibody (120024)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561840
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
TWSG1 may be involved in dorsoventral axis formation. Seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors. In addition to the anti-BMP function, also has pro-BMP activity, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. May be an important modulator of BMP-regulated cartilage development and chondrocyte differentiation. May play a role in thymocyte development.
Specifications
| TSG | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9EP52 | |
| Twsg1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSG, Cys25-Phe222. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 120024 | |
| Unconjugated | |
| Twsg1 | |
| 1810013J15Rik; 9030422N06Rik; AW552143; D17Ertd403e; PSEC0250; Tsg; Twg; twisted gastrulation BMP signaling modulator 1; twisted gastrulation homolog 1; twisted gastrulation homolog 1 (Drosophila); twisted gastrulation protein homolog 1; Twsg1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 65960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction