missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSGA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-89839
This item is not returnable.
View return policy
Description
TSGA2 Polyclonal specifically detects TSGA2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TSGA2 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Cancer/testis antigen 79, CT79, FLJ32753, h-meichroacidin, Male meiotic metaphase chromosome-associated acidic protein, Meichroacidin, radial spoke head 1 homolog, radial spoke head 1 homolog (Chlamydomonas), RSP44, RSPH10A, testes specific A2 homolog, testes specific gene A2 homolog, testis specific A2 homolog, testis specific A2 homolog (mouse), Testis-specific gene A2 protein, TSA2MGC126568, TSGA2MGC141927 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RSPH1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TAELIHLNHRYQGKFLNKNPVGPGKYVFDVGCEQHGEYRLTDMERGEEEEEEELVTVVPKWKATQITELALWTPTL | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 89765 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction