missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPO Monoclonal Antibody (CL13358)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546545
This item is not returnable.
View return policy
Description
Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 81%.
Mitochondrial peripheral-type benzodiazepine receptor (PBR) is an indispensable element of the steroidogenic machinery, where it mediates the delivery of cholesterol to the inner mitochondrial side chain cleavage cytochrome P-450 upon ligand activation. PBR is composed of three subunits, an isoquinoline binding site, a voltage-dependent anion channel and an adenine nucleotide carrier. PBR is genetically conserved from bacteria to humans and in humans is widely expressed in peripheral organs, whereas in the brain, it is sparse and located mainly in glial cells. Peroxisome proliferator perfluordecanoic acid (PFDA) inhibits the Leydig cell steroidogenesis by affecting PBR mRNA stability, thus inhibiting PBR expression, cholesterol transport into the mitochondria and subsequent steroid formation. A cytoplasmic protein, PRAX-1 ( peripheral benzodiazepine receptor-associated protein 1), is found to specifically interact with PBR.
Specifications
| TSPO | |
| Monoclonal | |
| 0.97 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P30536 | |
| Tspo | |
| Synthetic Peptide. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL13358 | |
| Unconjugated | |
| Tspo | |
| Benzodiazepin receptor (peripheral); benzodiazepine peripheral binding site; benzodiazepine receptor, peripheral; BPBS; BZRP; DBI; IBP; isoquinoline-binding protein; MBR; mDRC; mitochondrial benzodiazepine receptor; OTTHUMP00000028891; PBR; PBS; peripheral-type benzodiazepine receptor; pk18; PKBS; PTBR; Ptbzr; PTBZR02; RATPTBZR02; translocator protein; translocator protein (18kDa); TSPO; Tspo1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 706 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction