missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPY1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41198
This item is not returnable.
View return policy
Description
TSPY1 Polyclonal specifically detects TSPY1 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| TSPY1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TSPY1 | |
| Antibody was raised against a 20 amino acid peptide near the carboxy terminus of human TSPY1L. The immunogen is located within the last 50 amino acids of TSPY1L. Amino Acid Squence: CRNPLQYYKRMKPPEEGTETS | |
| Affinity Purified | |
| RUO | |
| 7258 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA | |
| Cancer/testis antigen 78, CT78DYS14, testis specific protein, Y-linked, testis specific protein, Y-linked 1, testis-specific Y-encoded protein 1, TSPYpJA923 | |
| Rabbit | |
| 34 kDa | |
| 0.1 mg | |
| Primary | |
| At least three isoforms of TSPY1 are known to exist; this antibody will detect only TSPY1L. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction