missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer TST Indicator Strips, Box of 200 Strips

Product Code. 31453993
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
31453993 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31453993 Supplier Tuttnauer Supplier No. WTL1980068
  • ONLINE EXCLUSIVE PRICE
    59.48 €
    46.03 € / Pack of 200
    Save 13.45 €
    22% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Type 5 unique point integrator indicator used in steam sterilization processes for reliable cycle indication.

  • For steam sterilization applications requiring clear indication performance.
  • Includes 200 strips in a convenient format for routine use.

Specifications

Autoclavable Autoclavable
Color White
For Use With (Equipment) Autoclaves
Product Type Indicator Strip
Material Paper
Latex Free Yes
Quantity 200/Box
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.