missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 572.00 €
Specifications
| Antigen | TTC24 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18464322
|
Novus Biologicals
NBP1-90920-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392921
|
Novus Biologicals
NBP1-90920 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTC24 Polyclonal specifically detects TTC24 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| TTC24 | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 164118 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QVLEKSRRLAERSTGRRLLGHLYNDLGLGYSQLQLFPLAVEAFLQALPLCWVPGEQATVLRNLGMAHNALGNYQEAREFHQKAADLHGSVG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ20249, tetratricopeptide repeat domain 24, tetratricopeptide repeat protein 24 | |
| TTC24 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title