missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93516-0.1ml
This item is not returnable.
View return policy
Description
TTC3 Polyclonal antibody specifically detects TTC3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TTC3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DCRR1tetratricopeptide repeat protein 3 (TPR repeat protein D)10RNF105DKFZp686M0150, E3 ubiquitin-protein ligase TTC3, EC 6.3.2.-, Protein DCRR1, RING finger protein 105, tetratricopeptide repeat domain 3, Tetratricopeptide repeat protein 3, TPR repeat protein 3, TPR repeat protein D, TPRDI, TPRDII, TPRDTPRDIII | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1400-1700 of human TTC3 (NP_003307.3). IASETQILEGSLGISVKSHCSTGDAHTVLSESNRNDEHCGNSNNKCEVIPESTSAVTNIPHVQMVAIQVSWNIIHQEVNTEPYNPFEERQGEISRIEKEHQVLQDQLQEVYENYEQIKLKGLEETRDLEEKLKRHLEENKISKTELDWFLQDLEREIKKWQQEKKEIQERLKSLKKKIKKVSNASEMYTQKNDGKEKEHELHLDQSLEISNTLTNEKMKIEEYIKKGKEDYEESHQRAVAAEVSVLENWKESEVYKLQIMESQAEAFLKKLGLISRDPAAYPDMESDIRSWELFLSNVTKE | |
| 0.1 mL | |
| Zinc Finger | |
| 7267 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction