missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC39C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | TTC39C |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18217262
|
Novus Biologicals
NBP2-56001 |
100 μL |
624.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600219
|
Novus Biologicals
NBP2-56001-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
TTC39C Polyclonal specifically detects TTC39C in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Spezifikation
| TTC39C | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C18orf17, Chromosome 18 Open Reading Frame 17, HsT2697, Tetratricopeptide Repeat Domain 39C, TPR Repeat Protein 39C | |
| TTC39C | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 125488 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:INMLLNNGFRESDQLFKQYRNHSPLMSFGASFVSFLNAMMTFEEEKMQLACDDLKTTEKLCESEEAGVIETIKNKIKKNVDVRKSAPSMVDRLQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts