missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF-1/ NKX2-1 Antibody (TTF1/8123R), Novus Biologicals™
Rabbit Monoclonal Antibody
298.00 € - 530.00 €
Specifications
| Antigen | TTF-1 / NKX2-1 |
|---|---|
| Clone | TTF1/8123R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694774
|
Novus Biologicals
NBP3-20231-20ug |
20 μg |
298.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653085
|
Novus Biologicals
NBP3-20231-100ug |
100 μg |
530.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TTF-1 / NKX2-1 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Tumor Biomarkers | |
| 10mM PBS with 0.05% BSA | |
| 7080 | |
| IgG κ | |
| Protein A or G purified |
| TTF1/8123R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
| Recombinant fragment (around aa1-400) of human TTF-1/ NKX2-1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title