missing translation for 'onlineSavingsMsg'
Learn More

TTLL11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00 € - 539.00 €

Specifications

Antigen TTLL11
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18496431
View Documents
Novus Biologicals
NBP2-14495-25ul
25ul
280.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18106731
View Documents
Novus Biologicals
NBP2-14495
0.1 mL
539.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TTLL11 Polyclonal specifically detects TTLL11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TTLL11
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
158135
This antibody was developed against a recombinant protein corresponding to the amino acids: VRKITLSRAVRTMQNLFPEEYNFYPRSWILPDEFQLFVAQVQMVKDDDPSWKPTFIVKPDGGCQGDGIYLIKDPSDIRLAGTLQSRPAVVQEY
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Polyclonal
Rabbit
Human
bA244O19.1, C9orf20, RP11-429D3.1, TTLL11 tubulin tyrosine ligase-like family, member 11
TTLL11
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.