Learn More
TTYH3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24707-1-AP-20UL
This item is not returnable.
View return policy
Description
The TTYH3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TTYH3. This antibody recognizes human antigen. The TTYH3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TTYH3 | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| hTTY3, KIAA1691, Protein tweety homolog 3, TTYH3, tweety homolog 3 (Drosophila) | |
| 20 μL | |
| 80727 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |