Learn More
Invitrogen™ TUBA1B/TUBA4A Recombinant Rabbit Monoclonal Antibody (24GB2020)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557300
 View more versions of this product
Product Code. 30275288
  • 146.00 € / 20µL
Description
TUBA1B/TUBA4A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TUBA1B and TUBA4A are genes that encode for alpha-tubulin isoforms, essential components of the microtubule system that plays a critical role in maintaining cell structure, intracellular transport, and cell division. TUBA1B, also known as tubulin alpha-1b chain, is located on chromosome 12q13.12 and has been linked to various cancers due to its influence on microtubule dynamics and cellular proliferation. It is also associated with immune cell infiltration and modulation of immune checkpoints, making it significant in tumor growth and metastasis. TUBA4A, known as tubulin alpha-4A, is found on chromosome 2q35 and is linked to neurodevelopmental disorders and neurodegenerative diseases such as amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD). Mutations in TUBA4A can lead to defects in microtubule stability and function, emphasizing its role in neuronal health. Both genes highlight the critical role of microtubules in cellular and neurological functions, and their mutations can contribute to diverse pathological conditions, including cancer and neurodegenerative diseases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TUBA1B/TUBA4A
Recombinant Monoclonal
Unconjugated
TUBA1B
TUBA1B, TUBA4A
A synthesized peptide derived from human TUBA1B/4A (aa 33-47).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2020
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05213, P68363, P68366, P68368, Q5XIF6, Q6P9V9
Rabbit
Affinity chromatography
RUO
10376, 22143, 22145, 316531, 500929, 7277
-20°C