missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBGCP5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | TUBGCP5 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640512
|
Novus Biologicals
NBP2-94420-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651622
|
Novus Biologicals
NBP2-94420-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TUBGCP5 Polyclonal antibody specifically detects TUBGCP5 in Rat samples. It is validated for Western BlotSpecifications
| TUBGCP5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 114791 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| gamma-tubulin complex component 5, gamma-tubulin complex component GCP5, GCP5KIAA1899GCP-5, tubulin, gamma complex associated protein 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TUBGCP5 (NP_443135.3). KEIKTDAHYSILSLLLCLSDSPSNSSYVETPRNKEVEKKDDFDWGKYLMEDEEMDIGPYMDTPNWSEESEEENDQQPLSREDSGIQVDRTPLEEQDQNRKL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title