missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin Beta 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99382-100ul
This item is not returnable.
View return policy
Description
Tubulin Beta 4 Polyclonal antibody specifically detects Tubulin Beta 4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Tubulin Beta 4 | |
| Polyclonal | |
| Western Blot 1:2000-1:5000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| TUBB5beta-5, Tubulin 5 beta, tubulin beta-4 chain, tubulin, beta 4, tubulin, beta, 5 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Tubulin Beta 4. | |
| 100 μg | |
| Apoptosis, Cell Biology, Cellular Markers | |
| 10382 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction