Learn More
TUFM Antibody (CL2243) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44456
Product Code. 30292025
  • 604.00 € / 100µL
Description
TUFM Monoclonal antibody specifically detects TUFM in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TUFM
Monoclonal
LYOPH
Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000
EFTu, EF-Tu, EFTUCOXPD4, EF-TuMT, elongation factor Tu, mitochondrial, P43, Tu translation elongation factor, mitochondrial
This antibody was generated using a recombinant protein sequence of P49411, with the exact immunogen sequence remaining proprietary.
100 μL
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
IgG2a
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry
CL2243
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
7284
Human
Purified