missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUFM Antibody (CL2245) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36753PCP
This item is not returnable.
View return policy
Description
TUFM Monoclonal antibody specifically detects TUFM in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| TUFM | |
| Monoclonal | |
| PerCP | |
| EFTu, EF-Tu, EFTUCOXPD4, EF-TuMT, elongation factor Tu, mitochondrial, P43, Tu translation elongation factor, mitochondrial | |
| This antibody was developed using a recombinant protein derived from P49411, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL2245 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 7284 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction