missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Tumor-derived Exosome ELISA Kit (Colorimetric)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Assay Sensitivity | <0.35 μg of overall exosome or < 50 pg targeted exosomes protein |
| Immunoassay Kit Format | Colorimetric Detection Kit |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Human Plasma |
| Target Species | Human |
| For Use With (Application) | ELISA, Immunoassay |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. After reconstitution store at -70°C. |
| Synonym | Cancer exosome, Cancer-derived exosomes, EV detection, Exosome, exosome detection, Exosome enrichment, Exosome kit, Exosome quantification, Exosomes ELISA, extracellular vesicle detection, Kit for exosomes |
| Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction