missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tunicamycin, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
Description
Tunicamycin causes accumulation of unfolded proteins in cell endoplasmic reticulum (ER) and induces ER stress, and causes blocking of DNA synthesis and cell cycle arrest in G1 phase. Tunicamycin inhibits gram-positive bacteria, yeasts, fungi, and viruses and has anti-cancer activity.Tunicamycin increases exosome release in cervical cancer cells.
Specifications
Specifications
| Chemical Name or Material | Tunicamycin |
| Physical Form | Solid |
| CAS | 11089-65-9 |
| Color | Light Yellow |
| Quantity | 2 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C39H64N4O16 |
| For Use With (Application) | COVID-19-anti-virus |
| Solubility Information | DMSO : 40 mg/mL (Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1NC(N([C@H]2[C@H](O)[C@H](O)[C@@H](C(C[C@@H]3[C@H](O)[C@H](O)[C@@H](NC(/C=C/[(CH2) 8-11]C(C)C)=O)[C@H](O[C@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4NC(C)=O)O3)O)O2)C=C1)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction