missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody (MTW-1) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-09045-0.2mg
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Neutralization
Specifications
| TWEAK/TNFSF12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunofluorescence, Neutralization | |
| MTW-1 | |
| Unconjugated | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| This antibody was raised by immunising SD rats with CD8-mTWEAK, a fusion of the human CD8alpha extracellular region to murine TWEAK. | |
| 0.2 mg | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| 8742 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto