Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
BIOPHARMAGREENER_CHOICE
Product Code. 18088794 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Clear
Colorless
Green
Skirt Style:
Semi-skirted
Skirted
Product Type:
BioBased LoBind PCR Plates
BioBased PCR Plates
No. of Wells:
96
384
Unit Size:
Pack of 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Product Type No. of Wells Unit Size
18088794 Colorless Skirted BioBased PCR Plates 384 Pack of 25
17882588 Green Skirted BioBased PCR Plates 96 Pack of 25
17802598 Clear Skirted BioBased PCR Plates 96 Pack of 25
18098794 Colorless Semi-skirted BioBased PCR Plates 96 Pack of 25
18078794 Colorless Semi-skirted BioBased LoBind PCR Plates 96 Pack of 25
18018804 Colorless Skirted BioBased LoBind PCR Plates 96 Pack of 25
18008804 Colorless Skirted BioBased LoBind PCR Plates 384 Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 18088794 Supplier Eppendorf™ Supplier No. 0030531051
  • ONLINE EXCLUSIVE PRICE
    256.28 €
    228.00 € / Pack of 25
    Save 28.28 €
    11% Off
In Stock
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Discover the new twin.tec™ PCR Plates BioBased made of bio-based polymer (mass balance approach). These plates allow you to significantly reduce your consumable-related carbon footprint while maintaining identical technical performance.
  • Reduction of consumable-related carbon footprint in the lab
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
  • Available with barcode (upon request)
TRUSTED_SUSTAINABILITY
Specifica
Barcode Custom Barcoding Available
Product Line Eppendorf
Color Colorless
Product Type BioBased PCR Plates
Skirt Style Skirted
Material Polypropylene, Polycarbonate
No. of Wells 384
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Quantity 25 Pk.
Fit for Purpose PCR Ready
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.