missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
GREENER_CHOICE

Product Code. 10440378 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
White
Skirt Style:
Semi-skirted
Skirted
Unskirted
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
Volume (Metric) Well:
150 μL
250 μL
Unit Size:
Pack of 10
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Purity or Quality Grade Volume (Metric) Well Unit Size
10440378 Blue Unskirted PCR Clean (Non-sterile) 150 μL Pack of 20
10740920 White Unskirted PCR Clean (Non-sterile) 150 μL Pack of 20
15685367 White Skirted Forensic DNA Grade (Non-sterile) 150 μL Pack of 10
15695367 White Semi-skirted Forensic DNA Grade (Non-sterile) 250 μL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10440378 Supplier Eppendorf™ Supplier No. 0030132718
  • ONLINE EXCLUSIVE PRICE
    133.00 €
    124.00 € / Pack of 20
    Save 9.00 €
    6% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.

  • White wells for better reflection.
  • High mechanical stability.
  • Effective sealing by raised well rims.
  • Skirted and semi-skirted frames.
  • Optimal heat transfer due to reduced wall thickness.
  • Autoclavable (121°C, 20 min.).
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color Blue
Product Type PCR Plate
Skirt Style Unskirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) RT-PCR applications
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 20 Pk.
Rimmed Yes
Well Color White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.