missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ Trace PCR Plates - PCR Plates
BIOPHARMAGREENER_CHOICE

Product Code. 17841447 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Colorless
Crystal Blue
Fuchsia
Product Type:
Trace PCR Plate
Trace PCR Plate LoBind™
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type Unit Size
17841447 Crystal Blue Trace PCR Plate Pack of 25
17831447 Colorless Trace PCR Plate Pack of 25
17851447 Fuchsia Trace PCR Plate Pack of 25
18068794 Colorless Trace PCR Plate LoBind™ Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17841447 Supplier Eppendorf™ Supplier No. 0030129776
  • ONLINE EXCLUSIVE PRICE
    165.00 €
    154.00 € / Pack of 25
    Save 11.00 €
    6% Off
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The Eppendorf twin.tec™ Trace PCR Plates feature an OptiTrack™ matrix and optical guiding grid for effortless well orientation and laser-engraved lot numbers for enhanced traceability.

  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 5 Plates
Product Line Eppendorf
Color Crystal Blue
Product Type Trace PCR Plate
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 25 Pk.
Well Color Colorless
Fit for Purpose PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.