missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TWIST1 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149159
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
TWIST1 acts as a transcriptional regulator. It inhibits myogensis by sequestering E proteins, inhibiting trans-activation by MEF2, and inhibiting DNA-binding by MYOD1 through physical interaction. TWIST1 also represses proinflammatory cytokines such as TNFA and IL1B. TWIST also regulates cranial suture patterning and fusion and gene expression differentially depending on dimer composition. Mutations in the gene are implicated in Saethre-Chotzen syndrome, a disease characterized by coronal synostosis, brachycephaly, low front hairline, facial asymmetry, hypertelorism, broad halluces, and clinodactyly.
Specifications
| TWIST1 | |
| Polyclonal | |
| Unconjugated | |
| TWIST1 | |
| AA960487; ACS3; B-HLH DNA binding protein; bHLHa38; BPES2; BPES3; charlie chaplin; class A basic helix-loop-helix protein 38; CRS; CRS1; CSO; H-twist; M-twist; p ACS3; Pde; pdt; Pluridigite; polydactyly EMS; SCS; Ska>m10Jus>; Ska>sup>m10Jus>/sup>; Ska10; Twist; twist basic helix-loop-helix transcription factor 1; twist family bHLH transcription factor 1; twist gene homolog 1; twist homolog 1; TWIST homolog of drosophila; Twist1; twist-related protein 1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 7291 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q15672 | |
| TWIST1 | |
| E. coli-derived recombinant human Twist-1, Met1-His202. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction