missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TXLNA Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5149030

Product Code. 30535326

  • 429.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

May be involved in intracellular vesicle traffic and potentially in calcium-dependent exocytosis in neuroendocrine cells. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TXLNA
Polyclonal
Unconjugated
TXLNA
Alpha-taxilin; DKFZp451J0118; il 14; IL14; IL-14; interleukin 14; MGC118870; MGC118871; RP4-622L5.4; taxilin alpha; TXLN; TXLNA
Sheep
Antigen affinity chromatography
RUO
200081
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with trehalose and no preservative
P40222
TXLNA
E. coli-derived recombinant human alpha -Taxilin, Met1-Lys162.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.