missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNL2 Monoclonal Antibody (791431)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561841
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.
Spécification
| TXNL2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O76003 | |
| GLRX3 | |
| E. coli-derived recombinant human Glutaredoxin 3/GLRX3, Asn126-Lys294. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 791431 | |
| Unconjugated | |
| GLRX3 | |
| bA500G10.4; FLJ11864; Glrx3; GLRX4; glutaredoxin 3; glutaredoxin 4; glutaredoxin-3; GRX3; GRX4; HUSSY-22; Picot; PKC interacting cousin of thioredoxin; PKC-interacting cousin of thioredoxin; PKCq-interacting protein; PKC-theta-interacting protein; testicular tissue protein Li 75; thioredoxin-like 2; thioredoxin-like protein 2; TXNL2; TXNL3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10539 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu