missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tylosin tartrate

Product Code. 16504234 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 16504234

Brand: Thermo Scientific Chemicals 463070050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74610-55-2
Molecular Formula C50H83NO23
Molecular Weight (g/mol) 1066.20
InChI Key ICVKYYINQHWDLM-UHFFFAOYNA-N
IUPAC Name 2,3-dihydroxybutanedioic acid; 2-[6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-16-ethyl-4-hydroxy-15-{[(5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl)oxy]methyl}-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
SMILES OC(C(O)C(O)=O)C(O)=O.CCC1OC(=O)CC(O)C(C)C(OC2OC(C)C(OC3CC(C)(O)C(O)C(C)O3)C(C2O)N(C)C)C(CC=O)CC(C)C(=O)C=CC(C)=CC1COC1OC(C)C(O)C(OC)C1OC

Specifications

Type Tylosin Tartrate
Quantity 5 g
Solubility Information Solubility aq. soln.: ca 50 g/L
Formula Weight 1066.2
Chemical Name or Material Tylosin tartrate
compliance-icons
Product Identifier
  • Tylosin tartrate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Tylosin tartrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.