missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tylosin tartrate, MedChemExpress
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
missing translation for 'unitSize'
50mg
Description
Tylosin tartrate is a macrolide antibiotic found naturally as a fermentation product of Streptomyces fradiae. Tylosin tartrate exerts potent antimicrobial activity against Gram-positive bacteria. Tylosin tartrate is widely used as a feed additive for promoting animal growth. Tylosin tartrate is used for veterinary purposes against bacterial dysentery and respiratory diseases in poultry, pigs and cattle.
Specifications
Specifications
| Chemical Name or Material | Tylosin tartrate |
| Physical Form | Solid |
| CAS | 74610-55-2 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C50H83NO23 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : ≥ 100 mg/mL (93.79 mM) ∣DMSO : ≥ 100 mg/mL (93.79 mM) |
| SMILES | C[C@@H](O[C@@]1([H])O[C@H]([C@H]([C@@H](CC2=O)O)C)[C@H](C[C@H](C(/C=C/C(C)=C/[C@H](CO[C@H](O[C@H](C)[C@@H](O)[C@H]3OC)[C@@H]3OC)[C@@H](CC)O2)=O)C)CC=O)[C@H]([C@@H]([C@H]1O)N(C)C)O[C@@](O[C@@H](C)[C@@H]4O)([H])C[C@]4(O)C.O[C@@H](C(O)=O)[C@@H](O)C(O)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction