missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase Antibody (SPM360), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32905-0.02mg
This item is not returnable.
View return policy
Description
Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Tyrosinase | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| P14679 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| SPM360 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA) | |
| Recombinant tyrosinase protein (Uniprot: P14679 ) | |
| 0.02 mg | |
| Lipid and Metabolism | |
| 7299 | |
| Store at 4C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction