missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase-Related Protein-1 Rabbit, Clone: TYRP1/1564R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | Tyrosinase-Related Protein-1 |
|---|---|
| Clone | TYRP1/1564R |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15981038
|
EnquireBio
7306-RBM4-P0 |
20 μg | |||||||
|
15991038
|
EnquireBio
7306-RBM4-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Tyrosinase-Related Protein-1 | |
| 200μg/mL | |
| Monoclonal | |
| Rabbit | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 7306 | |
| Recombinant human full-length TYRP1 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 75kDa |
| TYRP1/1564R | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_000541 | |
| TYRP1 | |
| IgG κ | |
| Protein A/G Purified | |
| Cytoplasmic | |
| SK-MEL-23, SK-MEL-19, SK-MEL-30, SK-MEL-37 cells or Melanoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title