missing translation for 'onlineSavingsMsg'
Learn More

Tyrosol, TRC

Product Code. 30012783
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012783 5 g 5g
30006191 10 g 10g
17984445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012783 Supplier Toronto Research Chemicals Supplier No. TRCT9478005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-94-0
Molecular Formula C8 H10 O2
Molecular Weight (g/mol) 138.16
Synonym 2-(4-Hydroxyphenyl)ethanol, Metoprolol Succinate Imp. G (EP), Metoprolol Tartrate Imp. G (EP), Phenethyl alcohol, p-hydroxy- (7CI,8CI), Tyrosol (6CI), 4-Hydroxybenzeneethanol, 2-(4-Hydroxyphenyl)ethyl alcohol, 2-(p-Hydroxyphenyl)ethanol, 4-(2-Hydroxyethyl)phenol, 4-Hydroxyphenethyl alcohol, 4-Hydroxyphenylethanol, NSC 59876, Tyrosol C, p-(2-Hydroxyethyl)phenol, p-HPEA, p-Hydroxyphenethyl alcohol, p-Tyrosol, β-(4-Hydroxyphenyl)ethanol, β-(p-Hydroxyphenyl)ethanol
IUPAC Name 4-(2-hydroxyethyl)phenol
SMILES OCCc1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H10O2/c9-6-5-7-1-3-8(10)4-2-7/h1-4,9-10H,5-6H2
Formula Weight 138.0681
Percent Purity >95%
Chemical Name or Material Tyrosol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.