missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A23, 99%, Thermo Scientific Chemicals

Product Code. p-7041797
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 15483099

Brand: Thermo Scientific Alfa Aesar J60308.#0

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent inhibitor of EGF receptor kinase activity

Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

Solubility
Soluble in DMSO, ethanol, or DMF

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118409-57-7
Molecular Formula C10H6N2O2
Molecular Weight (g/mol) 186.17
MDL Number MFCD00133899
InChI Key VTJXFTPMFYAJJU-UHFFFAOYSA-N
Synonym tyrphostin 23, tyrphostin a23, 3,4-dihydroxybenzylidene malononitrile, tyrphostin ag18, tyrphostin rg50810, unii-rv0gcd31oj, 2-3,4-dihydroxyphenyl methylidene propanedinitrile, 2-3,4-dihydroxybenzylidene malononitrile, alpha-cyano-3,4-dihydroxy cinnamonitrile, propanedinitrile,2-3,4-dihydroxyphenyl methylene
PubChem CID 2052
IUPAC Name 2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
SMILES C1=CC(=C(C=C1C=C(C#N)C#N)O)O

Specifications

Color Brown to Yellow
Quantity 5 mg
Solubility Information Soluble in DMSO, ethanol, or DMF
Formula Weight 186.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tyrphostin A23

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.