missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin B46, 98+%, Thermo Scientific Chemicals

Product Code. 15424249
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424249 10 mg 10mg
15434249 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424249 Supplier Thermo Scientific Alfa Aesar Supplier No. J61704.MA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of EGF receptor kinase activity

Inhibitor of EGF receptor kinase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133550-34-2
Molecular Formula C19H18N2O3
Molecular Weight (g/mol) 322.364
MDL Number MFCD00209865
InChI Key GSQOBTOAOGXIFL-WJDWOHSUSA-N
Synonym tyrphostin b46, z-2-cyano-3-3,4-dihydroxyphenyl-n-3-phenylpropyl prop-2-enamide, n-3 inverted exclamation marka-phenylpropyl-3,4-dihydroxybenzylidenecyanoacetamide
PubChem CID 5353390
IUPAC Name (Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide
SMILES C1=CC=C(C=C1)CCCNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 322.36
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tyrphostin B46

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.