missing translation for 'onlineSavingsMsg'
Learn More

TZ9, MedChemExpress

Product Code. 16490028
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490028 5 mg 5mg
16453577 50 mg 50mg
16447558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490028 Supplier Medchem Express Supplier No. HY18643
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TZ9 is a novel inhibitor of Rad6 ubiquitin conjugating enzyme(E2 enzyme); inhibits MDA-MB-231 cell proliferation with IC50 of ∼6 uM.

TZ9 is a novel inhibitor of Rad6 ubiquitin conjugating enzyme(E2 enzyme)

Specifications

Chemical Name or Material TZ9
Physical Form Solid
CAS 1002789-86-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H14N6O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 40 mg/mL (109.19 mM)
SMILES O=C(OCC1=NC(N)=NC(NC2=CC=CC=C2)=N1)C3=CC=C([N+]([O-])=O)C=C3
Molecular Weight (g/mol) 366.33
Formula Weight 366.33
Percent Purity 99.26%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.