missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ub-K11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120621
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, RAW264.7, C6, Mouse brain, Mouse kidney.
Exists either covalently attached to another protein, or free (unanchored). When covalently bound, it is conjugated to target proteins via an isopeptide bond either as a monomer (monoubiquitin), a polymer linked via different Lys residues of the ubiquitin (polyubiquitin chains) or a linear polymer linked via the initiator Met of the ubiquitin (linear polyubiquitin chains). Polyubiquitin chains, when attached to a target protein, have different functions depending on the Lys residue of the ubiquitin that is linked: Lys-6-linked may be involved in DNA repair; Lys-11-linked is involved in ERAD (endoplasmic reticulum-associated degradation) and in cell-cycle regulation; Lys-29-linked is involved in lysosomal degradation; Lys-33-linked is involved in kinase modification; Lys-48-linked is involved in protein degradation via the proteasome; Lys-63-linked is involved in endocytosis, DNA-damage responses as well as in signaling processes leading to activation of the transcription factor NF-kappa-B. Linear polymer chains formed via attachment by the initiator Met lead to cell signaling. Ubiquitin is usually conjugated to Lys residues of target proteins, however, in rare cases, conjugation to Cys or Ser residues has been observed. When polyubiquitin is free (unanchored-polyubiquitin), it also has distinct roles, such as in activation of protein kinases, and in signaling. [Uniprot].
Specifications
| Ub-K11 | |
| Polyclonal | |
| Unconjugated | |
| UBB | |
| UBB | |
| A synthetic peptide corresponding to a sequence containing polyubiquitinated protein (K11 linkage). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Dot Blot, ELISA, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P0CG47, P0CG49, P0CG51 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192255, 22187, 7314 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction