missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH5a/UBE2D1 Antibody (SR1267), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 € - 472.00 €
Specifications
| Antigen | UbcH5a/UBE2D1 |
|---|---|
| Clone | SR1267 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682394
|
Novus Biologicals
NBP3-22574-100ul |
100 μg |
472.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686784
|
Novus Biologicals
NBP3-22574-50ul |
50 μg |
349.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UbcH5a/UBE2D1 Monoclonal antibody specifically detects UbcH5a/UBE2D1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| UbcH5a/UBE2D1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| E2(17)KB1, Stimulator of Fe transportUBC4/5 homolog, UBC4/5, UBC5A, UbcH5, UbcH5A, UBCH5AEC 6.3.2.19, UBCH5SFT, Ubiquitin carrier protein D1, ubiquitin-conjugating enzyme E2 D1, Ubiquitin-conjugating enzyme E2(17)KB 1, Ubiquitin-conjugating enzyme E2-17 kDa 1, ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D1 | |
| A synthesized peptide derived from human UbcH5a/UBE2D1 (Uniprot #: P51668) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR1267 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 7321 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title