missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH8/Ube2L6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00032-50ul
This item is not returnable.
View return policy
Description
UbcH8/Ube2L6 Polyclonal antibody specifically detects UbcH8/Ube2L6 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| UbcH8/Ube2L6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6 | |
| Produced in rabbits immunized with purified, recombinant Human UbcH8/Ube2L6 (Accession#: O14933-1; Met1-Ser153) | |
| 50 μg | |
| Stem Cell Markers | |
| 9246 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction