missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2D1 Recombinant Rabbit Monoclonal Antibody (24GB1900)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557304
This item is not returnable.
View return policy
Description
UBE2D1 Recombinant Monoclonal Antibody for Western Blot, Flow
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is closely related to a stimulator of iron transport (SFT), and is up-regulated in hereditary hemochromatosis. It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases.
Specifications
| UBE2D1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| UBE2D1 | |
| (E3-independent) E2 ubiquitin-conjugating enzyme D1; E2 ubiquitin-conjugating enzyme D1; E2(17)KB 1; E2(17)KB1; SFT; Stimulator of Fe transport; UBC 4/5; UBC4/5; UBC4/5 homolog; UBC5A; UBCH 5; UBCH 5A; UBCH5; UBCH5A; UBE2D1; ubiquitin; Ubiquitin carrier protein D1; ubiquitin conjugating enzyme E2 D1; ubiquitin conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2 D1; Ubiquitin-conjugating enzyme E2(17)KB 1; ubiquitin-conjugating enzyme E2-17 kDa 1; ubiquitin-conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 1, UBC4/5 homolog; ubiquitin-protein ligase D1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216080, 361831, 7321 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1900 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D3ZDK2, P51668, P61080 | |
| UBE2D1 | |
| A synthesized peptide derived from human UBE2D1 (aa 110-147). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction