missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2I/Ubc9 Antibody (SAIC-36A-9) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
244.00 € - 635.00 €
Specifications
| Antigen | UBE2I/Ubc9 |
|---|---|
| Clone | SAIC-36A-9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671834
|
Novus Biologicals
NBP3-20104-0.025mg |
0.025 mg |
244.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634154
|
Novus Biologicals
NBP3-20104-0.2mg |
0.2 mg |
635.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBE2I/Ubc9 Monoclonal antibody specifically detects UBE2I/Ubc9 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass SpectrometrySpecifications
| UBE2I/Ubc9 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C358B7.1, EC 6.3.2, EC 6.3.2.-, P18, SUMO-1-protein ligase, SUMO-protein ligase, UBC9SUMO-conjugating enzyme UBC9, UBCE9, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, ubiquitin conjugating enzyme 9, Ubiquitin-conjugating enzyme E2 I, ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9), ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast), ubiquitin-conjugating enzyme UbcE2A, ubiquitin-like protein SUMO-1 conjugating enzyme, ubiquitin-protein ligase E2I, Ubiquitin-protein ligase I | |
| Peptide 'GLFIIDGK', derived from the SUMO-conjugating enzyme UBC9, conjugated to KLH. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| SAIC-36A-9 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins | |
| PBS | |
| 7329 | |
| IgG κ | |
| Protein A purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel