missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XII Antibody (OTI2D12), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44694
This item is not returnable.
View return policy
Description
Factor XII Monoclonal antibody specifically detects Factor XII in Human samples. It is validated for Western Blot
Specifications
| Factor XII | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| OTI2D12 | |
| Western Blot 1:2000 | |
| beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, F12, HAE3, HAEX, HAF, HAF beta-factor XIIa part 2, Hageman factor | |
| Human recombinant protein fragment corresponding to amino acids 73-372 of human F12(NP_000496) produced in E.coli. | |
| 100 μg | |
| Cardiovascular Biology | |
| 2161 | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction