missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE4B Recombinant Rabbit Monoclonal Antibody (24GB1765)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557825
This item is not returnable.
View return policy
Description
UBE4B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
Specifications
| UBE4B | |
| Recombinant Monoclonal | |
| Unconjugated | |
| UBE4B | |
| 4930551I19Rik; 4933406G05Rik; AU014668; D4Bwg0973e; E4; HDNB1; homologous to yeast UFD2; homozygously deleted in neuroblastoma 1; homozygously deleted in neuroblastoma-1; KIAA0684; mKIAA0684; RING-type E3 ubiquitin transferase E4 B; Ube4b; Ubiquitin conjugation factor E4 B; ubiquitin fusion degradation protein 2; ubiquitination factor E4B; ubiquitination factor E4B (homologous to yeast UFD2); ubiquitination factor E4B (UFD2 homolog, yeast); ubiquitination factor E4B, UFD2 homolog; ubiquitin-fusion degradation protein 2; UBOX3; UFD2; Ufd2a; UFD2A-III/UBE4B-III splice isoform; Ufd2p | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10277, 298652, 63958 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1765 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95155, Q9ES00 | |
| UBE4B | |
| A synthesized peptide derived from human UBE4B (aa 15-50). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction