missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ubiquilin 1 Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer's and Parkinson's disease.
Specifications
Specifications
| Antigen | Ubiquilin 1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | Ubqln1 |
| Gene Accession No. | Q9UMX0 |
| Gene Alias | 1110046H03Rik; 1810030E05Rik; AU019746; C77538; D13Ertd372e; Da41; Dsk2; hPLIC-1; I79_020705; Plic1; PLIC-1; Protein linking IAP with cytoskeleton 1; testicular tissue protein Li 219; ubiquilin 1; ubiquilin-1; UBQLN1; UBQN; XDRP1 |
| Gene Symbols | Ubqln1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?