missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ubiquilin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554231
This item is not returnable.
View return policy
Description
Immunogen sequence: ATEAPRLLLW FMPCLAGTGS VAGGIESRED PLMSEDPLPN PPPEVFPALD SAELGFLSPP FLHMLQDLVS TNPQQLQPEA HFQVQLEQL Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.
This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This gene is specifically expressed in the testis, and proposed to regulate cell-cycle progression during spermatogenesis.
Specifications
| Ubiquilin 3 | |
| Polyclonal | |
| Unconjugated | |
| UBQLN3 | |
| 4933400K24Rik; testicular tissue protein Li 220; TUP-1; ubiquilin 3; ubiquilin-3; Ubqln3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50613 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H347 | |
| UBQLN3 | |
| Recombinant protein corresponding to Human Ubiquilin 3. Recombinant protein control fragment (Product #RP-91762). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction