missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBTD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15705351 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15705351 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15705351 Supplier Invitrogen™ Supplier No. PA557450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TVFHMKRRLH AAEGVEPGSQ RWFFSGRPLT DKMKFEELKI PKDYVVQVIV SQPVQNPTPV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

UBTD2 gene ontology (GO) annotation includes cytoplasm and cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBTD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBTD2
Gene Accession No. Q8WUN7
Gene Alias 4930571L24; 9630054F20Rik; AI645720; DCUBP; DC-UbP; Dendritic cell-derived ubiquitin-like protein; RGD1305506; SB72; ubiquitin domain containing 2; ubiquitin domain-containing protein 2; Ubiquitin-like protein SB72; Ubtd2
Gene Symbols UBTD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBTD2. Recombinant protein control fragment (Product #RP-96924).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.