Learn More
UBXN2B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25141-1-AP-20UL
This item is not returnable.
View return policy
Description
The UBXN2B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UBXN2B. This antibody recognizes human, mouse antigen. The UBXN2B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UBXN2B | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| NSFL1 cofactor p37, p37, p97 cofactor p37, UBX domain protein 2B, UBXN2B | |
| 20 μL | |
| 137886 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |